360.55 GBP valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Use promo code "24075" to get your promotional price.
Battery Type | NiMH |
---|---|
Material | PTFE |
Pore Size | 0.2 μm |
Type | Pipette Filler |
For Use With (Application) | accurate pipetting |
For Use With (Equipment) | measuring, volumetric, and Pasteur pipets |
Functional Range | 0.1 to 200mL |
Includes | 230v charger with EURO plug, inductive charging and park station and standard wall bracket, pipet holder with safety valve and filter |
Motor Type | Pump Motor |
Plug Type | EU |
Weight (Metric) | 240 g |
Voltage | 240 V |