Thermo Scientific Chemicals D(+)-Biotin, 98%
CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol
Overview
68.64 GBP valid until 2025-08-31
Use promo code "24128" to get your promotional price.
Code: 29
Additional Details:
Additional Details: CAS Number: 58-85-5 Weight: 0.00100kg
Quantity 1 g
Packaging Glass Bottle
Disclaimers: RUO – Research Use Only
Product Code. 10043120
Quantity Price
1 £ 80.8 / 1g
In stock, ready to ship: 51
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Chemical Identifiers
CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
Synonym biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
PubChem CID 171548
ChEBI CHEBI:15956
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12
Specification
Chemical Name or Material D(+)-Biotin
CAS 58-85-5
Melting Point 231.0°C to 233.0°C
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C10H16N2O3S
MDL Number MFCD00005541
Quantity 1 g
Merck Index 15,1236
Specific Rotation + 89.00
Synonym biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
Synonym biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
Synonym biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
Synonym biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
Synonym biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
Synonym biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
Synonym biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
Synonym biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
Synonym biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
Synonym biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
Solubility Information Solubility in water: 0.22g/L (25°C). Other solubilities: more soluble in hot water and dil. alkali,0.8g/L alcohol,insoluble in most other common organic solvents
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
Molecular Weight (g/mol) 244.31
PubChem CID 171548
ChEBI CHEBI:15956
Formula Weight 244.31
Percent Purity 98%