Fisher Bioreagents D-Sucrose (Molecular Biology), Fisher BioReagents
For the preparation of density gradients for DNA ultracentrifugation
Overview
172.77 GBP valid until 2025-06-30
Use promo code "24201" to get your promotional price.
Code: 32
Additional Details:
Additional Details: CAS Number: 57-50-1 Weight: 10.05000kg
Quantity 10 kg
Packaging Poly Pail
Product Code. 10051413
Quantity Price
1 £ 252.0 / 10kg
Estimated Shipment
08-08-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Special Offers
Description and Specification
Chemical Identifiers
CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Specification
Alpha Vector Sucrose
Chemical Name or Material D-Sucrose
CAS 57-50-1
Melting Point 190°C
Color White
pH 6.5 to 7.5
Ignition Residue 0.01% max.
Loss on Drying 0.03% max.
Purity Grade Notes DNase-, RNase- and Protease-Free
Assay Percent Range ≥99.9%
Packaging Poly Pail
Molecular Formula C12H22O11
MDL Number MFCD00006626
Quantity 10 kg
Merck Index 15, 9012
Specific Rotation +66 to +67° (+ or -)
Synonym sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
Synonym sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
Synonym sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
Synonym sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
Synonym sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
Synonym sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
Synonym sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
Synonym sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
Synonym sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
Synonym sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
Solubility Information 0.005% max. Insoluble Matter
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 342.30
PubChem CID 5988
ChEBI CHEBI:17992
Formula Weight 342.29
Percent Purity ≥99.9%
Grade Molecular Biology
Physical Form Solid
D-Sucrose is suitable for the preparation of density gradients used in purification of proteins and nucleic acids by ultracentrifugation.
Alternative Products
Documentation Special Offers