| Chemical Name or Material |
D(+)-Biotin
|
| CAS |
58-85-5
|
| Melting Point |
231.0°C to 233.0°C
|
| Assay Percent Range |
98%
|
| Packaging |
Glass bottle
|
| Molecular Formula |
C10H16N2O3S
|
| MDL Number |
MFCD00005541
|
| Quantity |
500 mg
|
| Merck Index |
15,1236
|
| Specific Rotation |
+ 89.00
|
| Synonym |
biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
|
| Synonym |
biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
|
| Synonym |
biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
|
| Synonym |
biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
|
| Synonym |
biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
|
| Synonym |
biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
|
| Synonym |
biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
|
| Synonym |
biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
|
| Synonym |
biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
|
| Synonym |
biotin,d-biotin,vitamin h,coenzyme r,vitamin b7,bios ii,factor s,bioepiderm,d +-biotin,biodermatin
|
| Solubility Information |
Solubility in water: 0.22g/L (25°C). Other solubilities: more soluble in hot water and dil. alkali,0.8g/L alcohol,insoluble in most other common organic solvents
|
| InChI Key |
YBJHBAHKTGYVGT-UHFFFAOYNA-N
|
| SMILES |
OC(=O)CCCCC1SCC2NC(=O)NC12
|
| IUPAC Name |
5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
|
| Molecular Weight (g/mol) |
244.31
|
| PubChem CID |
171548
|
| ChEBI |
CHEBI:15956
|
| Formula Weight |
244.31
|
| Percent Purity |
98%
|