Thermo Scientific™ Nunc™ Cell Culture/Petri Dishes
Perform a variety of applications across a broad range of sizes and styles to suit a variety of experimental needs.
Overview
180.00 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Code: 76
Additional Details:
Additional Details: Weight: 5.40000kg
Disclaimers: Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Code. 10098720
Quantity Price
1 £ 270.0 / Pack of 80
In stock, ready to ship: 105
In stock, ready to ship: 2495
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Special Offers
Description and Specification
Specification
Closure Included Closure Included
Closure Material Polystyrene
Capacity (Metric) 35 mL (Well)
Culture Area 145 sq. cm
Gridded Not Gridded
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Nunclon Delta
Type Cell Culture Dish
Volume (Metric) Working 35 mL
Well Count 1
Includes Airvent
No. per Case 80
No. per Pack 10
Vent Lid with airvent
Height (Metric) 21 mm
Diameter (Metric) 150 mm
Features:
  • Top and bottom stacking rings for easier handling and more stable stacking
  • Sterilized by gamma irradiation
  • Available with or without air vents for gas exchange
  • 2 x 2mm grid available for cloning experiments and determination of plating efficiency
Ordering Information:
174926 and 169558 feature 2mm grids
171099 and 150340 for suspension culture - not Nunclon
Includes:
A certificate that assures user of biological performance as well as traceability.
Alternative Products
Documentation Special Offers