Thermo Scientific™ Extended Length 200μL ART™ Barrier Specialty Filter Pipettor Tips
Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Overview
78.88 GBP valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Code: 03
Additional Details:
Additional Details: Weight: 1.20600kg
Product Code. 10180621
Quantity Price
1 £ 211.0 / Pack of 768
Estimated Shipment
27-08-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Special Offers
Description and Specification
Specification
Description ART XLP 200, Filtered, Sterile,
Volume (Metric) 200 μL
Filtered Filtered
Sterility Sterile
Color Natural
Model ART XLP 200
Length (English) 3.59 in.
Length (Metric) 9.12cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Rack
Tip Style Extended Length
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile and PCR Ready
Features:
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
Designed to aid in pipetting for a variety of applications:
Extended length to prevent contamination by keeping the pipette shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson Pipetman™ ™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Alternative Products
Documentation Special Offers