Corning™ Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
Overview
Code: 66
Additional Details:
Additional Details: Weight: 6.77700kg
Product Code. 10327461
Quantity Price
1 £ 304.72 / Pack of 100
In stock, ready to ship: 10
In stock, ready to ship: 5
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Special Offers
Description and Specification
Specification
Format Standard
Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
For Use With (Application) Attachment and growth of anchorage-dependent cells
No. per Case 100/Cs.
No. per Pack 20/Pk.
Quantity 100/Cs.
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.