12.64 GBP valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Use promo code "24140" to get your promotional price.
| Quantity | 2.5 L |
|---|---|
| Packaging | Amber Glass Bottle |
| CAS | 141-78-6 |
|---|---|
| Molecular Formula | C4H8O2 |
| MDL Number | 9171 |
| InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
| PubChem CID | 8857 |
| ChEBI | CHEBI:27750 |
| IUPAC Name | ethyl acetate |
| Melting Point | -83.5°C |
|---|---|
| CAS | 141-78-6 |
| Color | Colorless |
| Boiling Point | 75°C |
| Quantity | 2.5 L |
| Packaging | Amber Glass Bottle |
| Physical Form | Liquid |
| Molecular Formula | C4H8O2 |
| MDL Number | 9171 |
| Vapor Pressure | 103mbar at 20°C |
| Viscosity | 0.45cp at 20°C |
| Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
| Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
| Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
| Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
| Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
| Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
| Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
| Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
| Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
| Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
| InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
| SMILES | CCOC(=O)C |
| IUPAC Name | ethyl acetate |
| Molecular Weight (g/mol) | 88.106 |
| PubChem CID | 8857 |
| ChEBI | CHEBI:27750 |
| Formula Weight | 88.11g/mol |
[Ethyl acetate]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]