Melting Point |
-83.5°C
|
CAS |
141-78-6
|
Color |
Colorless
|
Boiling Point |
75°C
|
Quantity |
5 L
|
Packaging |
HDPE Bottle
|
Physical Form |
Liquid
|
Molecular Formula |
C4H8O2
|
MDL Number |
9171
|
Vapor Pressure |
103mbar at 20°C
|
Viscosity |
0.45cp at 20°C
|
Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
InChI Key |
XEKOWRVHYACXOJ-UHFFFAOYSA-N
|
SMILES |
CCOC(=O)C
|
IUPAC Name |
ethyl acetate
|
Molecular Weight (g/mol) |
88.106
|
PubChem CID |
8857
|
ChEBI |
CHEBI:27750
|
Formula Weight |
88.11g/mol
|