Use promo code "24376" to get your promotional price.
Lithium aluminum Hydride, 2.4M Solution in THF, AcroSeal™
Use promo code "24376" to get your promotional price.
Specification
| Chemical Name or Material | Lithium Aluminum hydride |
|---|---|
| Color | Yellow |
| Name Note | 2.4M Solution in THF |
| CAS | 16853-85-3 |
| CAS | 109-99-9 |
| Molecular Formula | AlH4Li |
| Linear Formula | LiAlH4 |
| MDL Number | MFCD00011075 |
| UN Number | 1411 |
| Concentration | 9.5 to 10.5% (as LiAlH4) |
| Density | 0.9000g/mL |
| Merck Index | 15, 344 |
| Synonym | lithium aluminum hydride,lithium aluminum hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride |
| Synonym | lithium aluminum hydride,lithium aluminum hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride |
| Synonym | lithium aluminum hydride,lithium aluminum hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride |
| Synonym | lithium aluminum hydride,lithium aluminum hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride |
| Synonym | lithium aluminum hydride,lithium aluminum hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride |
| Synonym | lithium aluminum hydride,lithium aluminum hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride |
| Synonym | lithium aluminum hydride,lithium aluminum hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride |
| Synonym | lithium aluminum hydride,lithium aluminum hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride |
| Synonym | lithium aluminum hydride,lithium aluminum hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride |
| Synonym | lithium aluminum hydride,lithium aluminum hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride |
| Solubility Information | Solubility in water: vigorous reaction. |
| InChI Key | OCZDCIYGECBNKL-UHFFFAOYSA-N |
| SMILES | [Li+].[AlH4-] |
| IUPAC Name | lithium(1+) alumanuide |
| Molecular Weight (g/mol) | 37.95 |
| PubChem CID | 21226445 |
| Formula Weight | 37.95 |
| Percent Purity | 9.2 to 10.5% (as LiAlH4) |
| Packaging | AcroSeal™ Glass bottle |
| Specific Gravity | 0.9 |
| Quantity | 100 mL |
| Flash Point | −17°C |
| Physical Form | Viscous Liquid |
Technical documents
[Lithium aluminium hydride, 2.4M solution in THF]
[Danger]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}, {code=CARC. 2, sequence=2, description=Carcinogenicity Category 2}, {code=EYE DAM. 1, sequence=3, description=Serious eye damage/eye irritation Category 1}, {code=FLAM. LIQ. 2, sequence=4, description=Flammable liquid Category 2}, {code=SKIN CORR. 1A, sequence=5, description=Skin corrosion/irritation Category 1 A}, {code=STOT SE 3, sequence=6, description=Specific target organ toxicity Category 3}, {code=WATER-REACT. 1, sequence=7, description=Substance which\, in contact with water\, emits flammable gases Category 1}]
[{code=H318, sequence=1, description=Causes serious eye damage.}, {code=H314, sequence=2, description=Causes severe skin burns and eye damage.}, {code=H351, sequence=3, description=Suspected of causing cancer.}, {code=H335, sequence=4, description=May cause respiratory irritation.}, {code=H336, sequence=5, description=May cause drowsiness or dizziness.}, {code=H302, sequence=6, description=Harmful if swallowed.}, {code=H260, sequence=7, description=In contact with water releases flammable gases which may ignite spontaneously.}, {code=H225, sequence=8, description=Highly flammable liquid and vapour.}, {code=EUH019, sequence=91, description=May form explosive peroxides.}]
[{code=P210, sequence=50, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P231+P232, sequence=51, description=Handle under inert gas. Protect from moisture.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=54, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS07, sequence=3, description=EXCLAMATION MARK}, {code=GHS08, sequence=4, description=HEALTH HAZARD}]
Product Identifier
- Lithium aluminium hydride, 2.4M solution in THF
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H318- Causes serious eye damage.
- H314- Causes severe skin burns and eye damage.
- H351- Suspected of causing cancer.
- H335- May cause respiratory irritation.
- H336- May cause drowsiness or dizziness.
- H302- Harmful if swallowed.
- H260- In contact with water releases flammable gases which may ignite spontaneously.
- H225- Highly flammable liquid and vapour.
- EUH019- May form explosive peroxides.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232- Handle under inert gas. Protect from moisture.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.