151.84 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Color | Clear |
---|---|
Description | 3000 x G |
Capacity (Metric) | 5 mL |
Length (Metric) | 75 mm |
Material | Polypropylene |
Disposable | Single-use |
Packaging | 125/Pk., 1000/Cs. |
Sterility | Sterile |
Graduated | No |
Diameter (Metric) | 12 mm |
Closure Type | Not Included |
Product Type | Test Tube |