107.99 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Disposable | Single-use |
---|---|
Filter Type | Ultrafiltration |
For Use With (Equipment) | Centrifuge |
Material (Membrane) | PES |
MWCO | 30 kDa |
Sample Volume | 500 mL |
Sterility | Non-sterile |
Filtration Area | 64.5m m2 |
Housing Material | Polycarbonate |
Quantity | 25/Pk. |
Type | Centrifugal Concentrator |
Volume (Metric) | 0.5 mL |
Length (Metric) | 50 mm |