78.00 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Certifications/Compliance | Certified DNA/Rnase Free |
---|---|
Color | Assorted |
Volume (Metric) | 0.2 mL |
Quantity | 300 strips |
For Use With (Equipment) | Thermal Cyclers |
Product Line | MicroAmp™ |
Material | Polypropylene |
Content And Storage | Strip of 8 attached caps for use with 0.2 mL individual tubes, strip tubes (0.1 or 0.2 mL), or 48- or 96-well plates. Assorted colors. |
Shipping Condition | Room Temperature |
Closure Type | Domed Cap |
No. of Wells | 96 |
Format | 200 Strip Caps |
Product Type | 8-Strip Cap |