Code:
66
Corning™ CellBIND™ Surface Cell Culture Flasks
Increases suface wettability for more even and consistent cell attachment. Corning™ CellBIND™ Surface Cell Culture are available in a variety of sizes, designs and cap styles to meet all laboratory needs.
Additional Details:
Weight:
12.79129kg
Disclaimers:
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specification
Barcode | Yes |
---|---|
Capacity (English) | 8.45 oz. |
Closure Color | Orange |
Closure Material | HDPE |
Capacity (Metric) | 250 mL |
Material | Polystyrene |
For Use With (Application) | For optimal cell attachment |
Includes | Lot numbers for ease in traceability |
Neck Style | Angled |
No. per Case | 84/cs. |
Shape | Rectangular |
Surface Coating | Corning Cellbind |
Graduated | Yes |
Sterility | Sterile |
Closure Type | Vented |
Culture Area | 175 cm2 |
Volume (Metric) Working | 35 to 52.5 mL |