Use promo code "24112" to get your promotional price.
Invitrogen™ E-Gel™ 1 Kb Plus DNA Ladder
Powerful tool for estimating molecular weight of linear, double-stranded DNA fragments
136.67 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Code:
50
Additional Details:
Weight:
0.10000kg
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Includes:
E-Gel™ 1 Kb Plus DNA Ladder is supplied at 0.035μg/μL in 10mM Tris-HCl (pH 7.5), 1mM EDTA (pH 8.0), 0.0075% XCFF, 0.05% tartrazine, and 5% glycerol. Includes 500μL of the E-Gel 1 Kb Plus DNA Ladder, enough for 100 loads
Specification
Content And Storage |
• 2 x 1 mL E-Gel 1 Kb Plus DNA Ladder • 1 mL 1X E-Gel Sample Loading Buffer Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C. |
---|---|
Concentration | 0.025 μg/μL |
Gel Type | Agarose |
Product Type | DNA Ladder |
Ready to Load | Yes |
Sample Loading Volume | 1 mL |
Size Range | 100 to 15000 bp |
Volume (Metric) | 2 x 1000 μL |
Gel Compatibility | E-Gel™ agarose gels |
Green Features | Sustainable packaging |
Nucleic Acid Structure | Linear DNA |
Quantity | 100 applications |
Shipping Condition | Approved for shipment at Room Temperature or on Dry Ice |
Technology | Individual chromatography-purified DNA fragments |