Thermo Scientific™ TRACE™ TR-Wax GC Columns
Analyze a wide range of polar compounds with this low-bleed, general purpose, high polarity GC column.
Overview
647.18 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 16
Additional Details:
Additional Details: Weight: 0.10000kg
Disclaimers: Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Code. 10796324
Quantity Price
1 £ 653.0 / Each
In stock, ready to ship: 1
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Max. Temperature 240/260°C
Diameter (Metric) 0.53 mm
Diameter (Metric) Inner 0.53 mm
Product Line TRACE
Stationary Phase Trace™ TR-WAX
USP Type G16, G20
For Use With (Application) Aromatic Isomer Analysis,GC for Alcohols,GC for Ketones,GC for Glycols,GC for Aromatic Isomers
For Use With (Application) Aromatic Isomer Analysis,GC for Alcohols,GC for Ketones,GC for Glycols,GC for Aromatic Isomers
For Use With (Application) Aromatic Isomer Analysis,GC for Alcohols,GC for Ketones,GC for Glycols,GC for Aromatic Isomers
For Use With (Application) Aromatic Isomer Analysis,GC for Alcohols,GC for Ketones,GC for Glycols,GC for Aromatic Isomers
For Use With (Application) Aromatic Isomer Analysis,GC for Alcohols,GC for Ketones,GC for Glycols,GC for Aromatic Isomers
Film Thickness 1 μm
Length (Metric) 30 m
Polarity Polar
Description 30 m x 0.53 mm x 1 μm
Engineered for Superior Performance
  • Polar phase, polyethylene glycol
  • Highly cross-linked and fully deactivated
  • Solvent washable
Applications:
  • Esters
  • Alcohols
  • Ketones
  • Glycols
  • Aromatic isomers
Excellent Quality and Reproducibility:
Recommended Use:
  • Low-bleed even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance
  • Aldehydes
  • Essential Oils
Alternative Uses:
  • Acids
  • Alcohols
  • Alcohols in beverages
  • Alditol acetates (sugars)
  • Amines–Aliphatic
  • Amines–Aromatic
  • Herbicides
  • Ketones
  • Nitroaromatics
  • Phenols
  • Volatiles
Similar To:
  • DB-Wax
  • BP20
  • Rtx™-Wax
  • Stabilwax™
  • HP20M
  • HP-Wax
  • HP-INNOWax
  • SUPELCOWAX™ 10
  • AT™-Wax
  • Nukol™
  • CP Wax52CB
  • SUPEROX II
  • Carbowax™
  • PE-WAX
  • ZBWAX
Alternative Products