233.64 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Format | Standard |
---|---|
Lid | With Lid |
Culture Area | 9.5 cm2 |
Array | 2 x 3 |
Binding Type | Medium Binding |
Color | Clear |
Material | Polystyrene |
Volume (Metric) Well | 16.8 mL |
No. of Wells | 6 |
Volume (Metric) Working | 1.9 to 2.9 mL |
Sterility | Sterile |
Surface Treatment | Non-treated |
Well Shape | Flat Bottom (F-well) |
No. per Case | 100/Cs. |
No. per Pack | 5/Pk. |
Well Color | Clear |