Use promo code "25339" to get your promotional price.
Dichloromethane, 99+%, stab. with ca. 50ppm amylene
Use promo code "25339" to get your promotional price.
| Quantity | 2500 mL |
|---|
Chemical Identifiers
| CAS | 75-09-2 |
|---|---|
| Molecular Formula | CH2Cl2 |
| Molecular Weight (g/mol) | 84.93 |
| MDL Number | MFCD00000881 |
| InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| PubChem CID | 6344 |
| ChEBI | CHEBI:15767 |
| IUPAC Name | dichloromethane |
| SMILES | ClCCl |
Specification
| Chemical Name or Material | Dichloromethane |
|---|---|
| Melting Point | -95°C |
| Name Note | stabilized with ca. 50ppm amylene |
| CAS | 75-09-2 |
| Density | 1.325 |
| Boiling Point | 39°C to 40°C |
| Quantity | 2500 mL |
| Odor | Sweet |
| Refractive Index | 1.4244 |
| Molecular Formula | CH2Cl2 |
| MDL Number | MFCD00000881 |
| UN Number | UN1593 |
| Beilstein | 1730800 |
| Merck Index | 14,6063 |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Solubility Information | Miscible with ethyl acetate,alcohol,hexanes,methanol,diethyl ether,n-octanol,acetone benzene,carbon tetrachloride,diethyl ether and chloroform. Immiscible with water. |
| InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
| SMILES | ClCCl |
| IUPAC Name | dichloromethane |
| Molecular Weight (g/mol) | 84.93 |
| PubChem CID | 6344 |
| ChEBI | CHEBI:15767 |
| Formula Weight | 84.93 |
| Percent Purity | ≥99% |
Technical documents
[Dichloromethane]
[Warning]
[{code=H315, sequence=1, description=Causes skin irritation.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H336, sequence=3, description=May cause drowsiness or dizziness.}, {code=H351, sequence=4, description=Suspected of causing cancer.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=52, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P304+P340, sequence=53, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P312, sequence=54, description=Call a POISON CENTER/doctor/if you feel unwell.}, {code=P337+P313, sequence=55, description=If eye irritation persists: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Dichloromethane}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}, {code=GHS08, sequence=2, description=HEALTH HAZARD}]
Product Identifier
- Dichloromethane
Signal Word
- Warning
Hazard Statement
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
- H336- May cause drowsiness or dizziness.
- H351- Suspected of causing cancer.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312- Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313- If eye irritation persists: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contains : Dichloromethane