Use promo code "24112" to get your promotional price.
Invitrogen™ 1 Kb Plus DNA Ladder
Powerful tool for estimating molecular weight of linear, double-stranded DNA fragments
146.00 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Code:
50
Additional Details:
Weight:
1.00000kg
Requires:
Gel-loading buffer (Invitrogen part no. 10816015)
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specification
Content And Storage |
• 500 µL 1 Kb Plus DNA Ladder • 1 mL 10X BlueJuice Gel Loading Buffer Store at -20°C. |
---|---|
Concentration | 0.5 μg/μL |
Gel Type | Agarose |
Product Type | DNA Ladder |
Ready to Load | No |
Sample Loading Volume | 1 mL |
Size Range | 100 to 15000 bp |
Volume (Metric) | 500 μL |
Gel Compatibility | Agarose gel |
Green Features | Sustainable packaging |
Kit Contents | 1 μL DNA Ladder (500 ng), 1 μL Sample Loading Buffer, 500 Applications |
No. of Reactions | 500 Applications |
Quantity | 500 Applications |
Shipping Condition | Approved for shipment at Room Temperature or on Dry Ice |
Technology | Individual chromatography-purified DNA fragments |