19.02 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Capacity | 500 mL |
---|---|
Content And Storage | 15°to 30°C |
Dilution | 10 X |
Product Type | Phosphate Buffered Saline (PBS) |
Quantity | 500mL |
For Use With (Application) | Mammalian Cell Culture |
Osmolality | 2800 to 3100 mOsm/kg |
pH | 7.3 to 7.5 |
Regulatory Status | For in-vitro diagnostic use |
Form | Liquid |