Use promo code "25339" to get your promotional price.
Sartorius Microsart™ @Filter 250 in Bags
Membrane filters suitable for use in the Microsart @Filter units
254.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Code:
NEW
Additional Details:
Weight:
0.01000kg
Specification
Color | Gray |
---|---|
Disposable | Single-use |
Format | Disc |
Material | Cellulose Nitrate |
Material (Membrane) | Cellulose Nitrate |
Pore Size | 0.7 μm |
Sample Volume | 250 mL |
Sterility | Sterile |
Grid Color | White |
Type | Filter Tip |
Diameter (Metric) | 47 mm |