77.20 GBP valid until 2025-12-31
Use promo code "24081" to get your promotional price.
Use promo code "24081" to get your promotional price.
Ensure accurate, precise pipetting. Finntip™ Specific Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Description | Finntip 10, Non-Filtered, Non-sterile, Rack |
---|---|
Volume (Metric) | 10 μL |
Filtered | Non-filtered |
Sterility | Non-sterile |
Color | Pink |
Model | Finntip 10 |
Length (English) | 1.2 in. |
Length (Metric) | 3.2cm |
Format | Rack |
Color Code | Pink |
For Use With (Equipment) | Thermo Scientific Finnpipette 10 |
Product Line | FinnTip |
Fit for Purpose | Certified Quality |