191.83 GBP valid until 2025-07-31
Use promo code "24075" to get your promotional price.
Use promo code "24075" to get your promotional price.
Accuracy | ±2.5 to 1.0% |
---|---|
Compatible Tips | Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10 |
Description | Finnpipette F3 1-10μL micro |
Color | Pink |
Model | F3 |
Number of Channels | 1 |
Product Line | Finnpipette™ F3 |
Type | Variable Volume Pipette |
Volume (Metric) | 1 to 10 μL |
Increments | 0.02μL |
Precision | 2.0 to 0.5% |
Color Code | Pink |