53.82 GBP valid until 2025-12-31
Use promo code "24310" to get your promotional price.
Use promo code "24310" to get your promotional price.
The Thermo Scientific™ Finntip™ Flex™ Pipette Tips reduce the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI). Ensure a better seal with the pipetter module, for higher accuracy and precision.
Description | Finntip Flex 10, Non-Filtered, Non-sterile, Bulk |
---|---|
Volume (Metric) | 10 μL |
Filtered | Non-filtered |
Sterility | Non-sterile |
Color | Pink |
Model | Finntip Flex 10 |
Length (English) | 1.1 in. |
Length (Metric) | 3.3cm |
Format | Bulk |
Packaging | 1000/Bag |
Color Code | Pink |
For Use With (Equipment) | Thermo Scientific Finnpipette |
Product Line | FinnTip Flex |
Fit for Purpose | Certified Quality |