Epredia™ SuperFrost Ultra Plus™ GOLD Adhesion Slides
Epredia™TM SuperFrost Plus™ Gold slides are recommended for hard-to-hold tissue samples like bone, brain, breast and skin. They are ideal for special stains, immunocytochemical and in-situ DNA hybridization on these tissue types.
Overview
131.10 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Code: 03
Additional Details:
Additional Details: Weight: 0.32400kg
Product Code. 11976299
Quantity Price
1 £ 141.45 / Pack of 72
In stock, ready to ship: 127
In stock, ready to ship: 2000
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Length (Metric) 75 mm
Width (Metric) 25 mm
Color White
  • No time wasted on slide-coating proceduress
  • No money wasted on expensive adhesives
  • Reliable and uniform coating
  • Minimal sample loss
  • Reliable and safe adhesive action