Invitrogen™ UltraPure™ DNase/RNase-Free Distilled Water
UltraPure™ DNase/RNase-Free Distilled Water
Overview
123.33 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Code: 50
Additional Details:
Additional Details: CAS Number: 7732-18-5 Weight: 5.00000kg
Quantity 10 x 500 mL
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Product Code. 12060346
Quantity Price
1 £ 185.0 / Pack of 10
In stock, ready to ship: 920
Add to basket
For Research Use Only. All usage must comply with product instructions.
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Packaging 1 Each
Treatment(s) Not DEPC-treated
Quantity 10 x 500 mL
UltraPure™ DNase/RNase-Free Distilled Water is designed for use in all molecular biology applications. It is 0.1-μm membrane-filtered and tested for DNase and RNase activity.
Performance and Quality Testing: No DNase, RNase, or protease activity detected. Our distilled water system is routinely monitored to show compliance with current USP monograph test requirements for Water for Injection (WFI).