140.25 GBP valid until 2025-07-31
Use promo code "24075" to get your promotional price.
Use promo code "24075" to get your promotional price.
Accuracy | ±0.025, ±0.075, ±0.1μL |
---|---|
Compatible Tips | D10, DL10, DF10ST, DFL10ST tip |
Adjustable Tip Spacing | No |
Autoclavable | Autoclavable |
Color | Red |
Dispensing Speed | Manual |
Disposable | Reusable |
For Use With | D-10, DL10, DF10ST, DFL10ST Tip |
Grip Type | Ergonomic |
Material | Polypropylene |
Model | P10L |
Number of Channels | 1 |
Product Line | Pipetman |
Programmability | No |
Sterility | Non-sterile |
Type | Mechanical Pipette |
Volume (Metric) | 1 to 10 μL |
Finger Style | Ergonomic |
For Use With (Application) | Transporting a measured volume of liquid |
For Use With (Equipment) | D10, DL10, DF10ST, DFL10ST tip |
Includes | Stainless Steel Tip Ejector |
Quantity | 1 |
Warranty | 1 Year |
Length (English) | 12.6 in. |
Length (Metric) | 32 cm |
Width (English) | 4.33 in. |
Width (Metric) | 11 cm |
Height (English) | 1.57 in. |
Height (Metric) | 4 cm |
Color Code | Red |
Fit for Purpose | Certified Quality |