Gibco™ DPBS, no calcium, no magnesium
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
Overview
102.64 GBP valid until 2025-12-31
Use promo code "24341" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: Weight: 0.01100kg
Disclaimers: For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Product Code. 12559069
Quantity Price
1 £ 127.0 / Pack of 10
In stock, ready to ship: 1689
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Additive No Sodium Pyruvate
Classification Animal Origin-Free
Color Clear
Content And Storage Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 36 months from date of manufacture
Packaging Type Bottle
Dilution 1 X
Product Line Gibco™
Product Type DPBS
Quantity 10 x 500 mL
Solution Type Dulbecco's Phosphate Buffered Saline
For Use With (Application) Mammalian Cell Culture
Osmolality 270 - 300 mOsm/kg
pH 7.0 to 7.3
Phenol Red Indicator No Phenol Red
Shipping Condition Room Temperature
Sterility Sterile-filtered
Form Liquid
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red