204.99 GBP valid until 2025-08-22
Use promo code "24148" to get your promotional price.
Use promo code "24148" to get your promotional price.
Volume (Metric) | 25 mL |
---|---|
Sterility | Sterile |
Color | Colorless |
Autoclavable | Not Autoclavable |
Clean Claims | Biopur™ (Sterile) |
Format | Individually Wrapped |
Tip Style | Extended Length,Positive Displacement |
Tip Style | Extended Length,Positive Displacement |
Packaging | 100 Pk., Individually Blister-wrapped |
Product Type | Pipette Tips |
Color Code | Red |
Disposable | Single-use |
For Use With (Equipment) | Eppendorf Multipette |
Graduated | Yes |
Product Line | CombiTips Advanced |
Material | Polypropylene |
Fit for Purpose | Sterile and PCR Ready |