Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Obtain a comprehensive solution when plastic is preferred to fiberboard.
Additional Details:
Weight:
1.74600kg
Specification
Type | Cryogenic Storage Box |
---|---|
Capacity (English) | 5.29 cu. ft. |
Capacity (Metric) | 1.5 mL |
For Use With (Equipment) | Locator Jr. Plus, Locator 4 Plus, Locator 6 Plus |
Height (English) Exterior | 2 in. |
Vial Capacity | 100 |
Holds | 1 and 1.5 mL Vials |
Quantity | 10/Cs. |