79.07 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Product Type | Protective Coveralls |
---|---|
Size | 3X-Large |
Gender | Unisex |
Material | SMS Polypropylene |
Color | White |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Hood Type | 2-panel |
Closure Type | Front Zipper with Storm Flap |
Seam Type | Serged |
Sterility | Non-sterile |
Protection Class | PPE Directive Category III, Type 5/6 |
Model | 4515W |
Size (English) Chest | 49 to 52 in. |
Size (Metric) Chest | 124 to 132 cm |
Height (English) | 76 to 78 in. |
Height (Metric) | 194 to 200 cm |