SGE™ BP20 (WAX) Capillary Columns
Industry standard WAX columns
Overview
Code: 13
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 12995581
Quantity Price
1 £ 627.0 / Each
Estimated Shipment
12-09-2025
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Diameter (Metric) Inner 0.53 mm
Stationary Phase SGE™ BP20 (Wax)
For Use With (Application) Ketones Analysis,Ketones,Aldehydes Analysis,Fragrance Analysis
For Use With (Application) Ketones Analysis,Ketones,Aldehydes Analysis,Fragrance Analysis
For Use With (Application) Ketones Analysis,Ketones,Aldehydes Analysis,Fragrance Analysis
For Use With (Application) Ketones Analysis,Ketones,Aldehydes Analysis,Fragrance Analysis
Particle Size 1 μm
Film Thickness 1 μm
Length (Metric) 30 m
Polarity Polar
  •  Very polar polyethylene glycol (PEG) phase
  • Cross-linked for stability and washing
  • Also ideal for xylene and aromatic isomer
  • 0.1 to 1μm film thickness: 20° to 260/280°C
Alternative Products