Waters Corporation Atlantis HILIC Column
HILIC separation mechanisms provide orthogonal analyte selectivity compared to traditional chromatographic reversed-phase approaches.
Additional Details:
Weight:
0.50000kg
Specification
Diameter (Metric) | 3 mm |
---|---|
Length (Metric) | 50 mm |
Max. Pressure | 6000 psi (415 Bar) |
Particle Shape | Spherical |
Particle Size | 5 μm |
Pore Size | 100 Å |
Quantity | 1/Ea. |
USP Type | L3 |
Endcapped | No |
Type | Column |