108.20 GBP valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Use promo code "25076" to get your promotional price.
Description | ART Barrier, Filtered, Sterile, Hinged Rack |
---|---|
Volume (Metric) | 200 μL |
Filtered | Filtered |
Sterility | Sterile |
Color | Natural |
Model | ART Barrier |
Length (English) | 3.6 in. |
Length (Metric) | 9.09 cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Hinged Rack |
Tip Style | Low Retention |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Fit for Purpose | Sterile and PCR Ready |