Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls
Use for the best results with fluorescence Western blotting detection applications. Cytiva Life Sciences™ Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Overview
222.74 GBP valid until 2025-12-31
Use promo code "24363" to get your promotional price.
Code: GH
Additional Details:
Additional Details: CAS Number: 9004-70-0 Weight: 0.27000kg
Product Code. 15229804
Quantity Price
1 £ 260.0 / 1 roll
In stock, ready to ship: 29
Add to basket
  • Description and Specification
  • Alternative Products
  • Safety and Handling
Description and Specification
Specification
Length (Metric) 4 m
Width (Metric) 20 cm
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Format Roll
Material 100% Pure Nitrocellulose
Pore Size 0.2 μm
Supplied in rolls.
  • Low background enables a dynamic range for complicated samples
  • Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
Alternative Products
Safety and Handling
[Amersham Protran Premium]
[Warning]
[{code=FLAM. SOL. 2, sequence=1, description=Flammable solid Category 2}]
[{code=H228, sequence=1, description=Flammable solid.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P210, sequence=52, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}]
[{code=GHS02, sequence=1, description=FLAME}]
Product Identifier
  • Amersham Protran Premium
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228- Flammable solid.
Precautionary Statement
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.