15.88 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Certifications/Compliance | ANSI Z87.1-2015, CSA Z94.3 |
---|---|
Lens Coating | Anti-fog |
Lens Tint | Clear |
Product Type | Safety Goggles |
Frame Color | Red |
Model | GG501SGAF |
Ventilation Details | Vented |
Material | Polycarbonate |