Use promo code "25339" to get your promotional price.
Dichloromethane, 99+%, extra pure, stabilized with ethanol
Use promo code "25339" to get your promotional price.
| Quantity | 5 L |
|---|---|
| Packaging | Plastic Bottle |
Chemical Identifiers
| CAS | 75-09-2 |
|---|---|
| Molecular Formula | CH2Cl2 |
| Molecular Weight (g/mol) | 84.93 |
| MDL Number | MFCD00000881 |
| InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| PubChem CID | 6344 |
| ChEBI | CHEBI:15767 |
| IUPAC Name | dichloromethane |
| SMILES | ClCCl |
Specification
| Chemical Name or Material | Dichloromethane |
|---|---|
| CAS | 75-09-2 |
| CAS | 64-17-5 |
| Density | 1.3250g/mL |
| Quantity | 5 L |
| Packaging | Plastic Bottle |
| Physical Form | Liquid |
| Assay Percent Range | 99+% |
| Residue after Evaporation | 0.05% max. |
| Molecular Formula | CH2Cl2 |
| Linear Formula | CH2Cl2 |
| MDL Number | MFCD00000881 |
| Beilstein | 01, 60 |
| Water | 0.1% max. (K.F.) |
| Fieser | 01,676; 02,273; 04,337; 07,239; 13,106 |
| Merck Index | 15, 6135 |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
| InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
| SMILES | ClCCl |
| IUPAC Name | dichloromethane |
| Molecular Weight (g/mol) | 84.93 |
| PubChem CID | 6344 |
| ChEBI | CHEBI:15767 |
| Formula Weight | 84.93 |
| Percent Purity | ≥99.0% |
| Grade | Extra Pure |
Technical documents
[Dichloromethane, stabilized with ethanol]
[Warning]
[{code=SKIN IRRIT. 2, sequence=1, description=Skin corrosion/irritation Category 2}, {code=EYE DAM. 2, sequence=2, description=Serious eye damage/eye irritation Category 2}, {code=CARC. 2, sequence=3, description=Carcinogenicity Category 2}, {code=STOT SE 3, sequence=4, description=Specific target organ toxicity Category 3}]
[{code=H315, sequence=1, description=Causes skin irritation.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H336, sequence=3, description=May cause drowsiness or dizziness.}, {code=H351, sequence=4, description=Suspected of causing cancer.}]
[{code=P202, sequence=51, description=Do not handle until all safety precautions have been read and understood.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=53, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P308+P313, sequence=55, description=IF exposed or concerned: Get medical advice/attention.}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}, {code=GHS08, sequence=2, description=HEALTH HAZARD}]
Product Identifier
- Dichloromethane, stabilized with ethanol
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
- H336- May cause drowsiness or dizziness.
- H351- Suspected of causing cancer.
Precautionary Statement
- P202- Do not handle until all safety precautions have been read and understood.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313- IF exposed or concerned: Get medical advice/attention.