17.48 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Certifications/Compliance | ASTM D3577, TYPE II, PPE 89/686, EN 420, 388 & 374 CATEGORY III |
---|---|
Color | Green,Blue |
Color | Green,Blue |
Disposable | Single-use |
Hand | Ambidextrous |
Material | Nitrile,Neoprene |
Material | Nitrile,Neoprene |
Powdered | Powder Free |
Product Type | Disposable Gloves |
Size | X-Large |
Thickness (Metric) | 0.198 mm |
Finger Style | Textured |
Glove Cuff Style | Extended, Beaded |
Model | 93-260 |
Packaging Type | Dispenser box |
Puncture Rating | 0.65 AQL |
Quantity | 50Pack |
Resistant To | Harsh chemicals including acids, solvents and bases |
Tensile Strength | 22 MPa |
Length (Metric) | 300 mm |
Width (Metric) | 116 mm |