164.67 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Content And Storage |
• 2 x 1 mL E-Gel Ultra Low Range DNA Ladder • 1.25 mL 1X E-Gel Sample Loading Buffer Store at room temperature or at 4°C for up to 6 months. For longer periods store at -20°C. |
---|---|
Concentration | 0.025 μg/μL |
Gel Type | Agarose |
Product Type | DNA Ladder |
Ready to Load | Yes |
Sample Loading Volume | 1.25 mL |
Size Range | 10 to 300 bp |
Volume (Metric) | 2 x 1000 μL |
Gel Compatibility | Double Comb E-Gels, Single Comb E-Gels |
Green Features | Sustainable packaging |
Quantity | 100 applications |
Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
Technology | Individual chromatography-purified DNA fragments |