Thermo Scientific™ Nunc™ EasYDish™ Dishes
Next generation cell culture dishes that improve handling, stacking and transporting of cell cultures in your lab.
Overview
166.14 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Additional Details:
Additional Details: Weight: 5.61600kg
Product Code. 15893937
Quantity Price
1 £ 277.0 / Pack of 240
In stock, ready to ship: 73
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Certifications/Compliance Nunclon Delta certification
Closure Included Closure Included
Closure Material Polystyrene
Description Nunc EasYDish
Autoclavable Not Autoclavable
Bottom Shape Flat
Capacity (Metric) 12.5 mL (Well)
Color Clear
Culture Area 56.7 sq. cm
Disposable Reusable
Gridded Not Gridded
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Nunclon Delta
Type Cell Culture Dish
Volume (Metric) Working 12.5 mL
Well Count 1
For Use With (Application) Cell Culture
Includes Lid
No. per Case 240
No. per Pack 10
Temperature Range (Metric) 37°C
Height (Metric) 21 mm
Diameter (Metric) 100 mm
Beveled Grip Ring
  • Ensures lid stays in position when dish is being handled
  • Makes it easier to grasp and manage with a gloved hand
Proprietary Notch Design
  • Maximizes ventilation while maintaining sterility
  • Restricts the movement of the lid ensuring secure lid-to-dish fitting
  • Increases the physical strength of the dish
Raised Outer Edge on Dish Lid
  • Allows stabled stacking during transportation of cell cultures
Orientation Marks on Dish Bottom
  • Simplifies locating of cells under the microscope
Writing Area
  • Improves traceability of cell cultures
Re-Sealable Packaging
  • Promotes cleanliness of opened packs
Specifications:
  • Material: Polystyrene, USP VI
  • Contaminant Free: RNase/DNase free, Human DNA free, non-cytotoxic, non-pyrogenic
  • Sterility: Gamma Irradiation to SAL of 10-6
  • Shelf Life: 5 years from manufacturing date
  • Certificate of Quality in every case
  • Surface Treatment: Nunclon Delta for adherent cell cultures