717.36 GBP valid until 2025-07-31
Use promo code "24075" to get your promotional price.
Use promo code "24075" to get your promotional price.
Calibration | External |
---|---|
Certifications/Compliance | GLP with optional printer compliance |
Connection Type | RS232 |
Capacity | 220 g |
Display Type | Backlit LCD |
Readability | 0.0001 g |
Includes | AC adapter |
Linearity | 0.0002 g |
Min. Sample Weight | 200 mg |
Model | FAS224/E |
Repeatability | 0.0001 g |
Stabilization Time | 4 sec. |
Type | Analytical Balance |
Warranty | 2 Years |
Weighing Modes | Weighing, Parts Counting, Percent Weighing |
Dimensions (D x W x H) | 317 x 201 x 303 mm |
Diameter (Metric) Outer Pan | 90 mm |
Electrical Requirements | 100/240 V ∼200 mA 50/60 Hz 12/18 VA |
Amperage | 0.5 A |
Frequency | 50/60 Hz |