Axygen™ Axypet™ Pro Single Channel Pipettor
Corning™ Axygen™ Axypet™ Pro Pipettors are engineered to provide the highest levels of comfort, accuracy, and precision.
Additional Details:
Weight:
0.23000kg
Specification
| Accuracy | ±3.5 to ±0.8 |
|---|---|
| Certifications/Compliance | ISO8655 |
| Autoclavable | Autoclavable |
| Color | Yellow |
| Grip Type | Ergonomic |
| Number of Channels | 1 |
| Product Line | Axypet Pro |
| Type | Pipette |
| Volume (Metric) | 2 to 20 μL |
| Finger Style | Ergonomic |
| Includes | Instruction manual, QC Certificate, In-lab calibration tool |
| Increments | 0.02 μL |
| Precision | ≤1.5 to ≤0.3 |
| Quantity | 1 |
| Warranty | 3 months |