50.80 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Use promo code "24135" to get your promotional price.
Volume (Metric) | 200 μL |
---|---|
Filtered | Filtered |
Sterility | Sterile |
Color | Natural |
Rack Color | Blue |
Autoclavable | Not Autoclavable |
Precision | 0.2 to 0.7% |
Accuracy | 0.6 to 1.8% |
Length (English) | 2.3 in. |
Length (Metric) | 58 mm |
Certifications/Compliance | Certificate of Sterility, COA |
Clean Claims | Certified Free of: Rnase/Dnase, DNA, Pyrogen, Endotoxin, Bioburden, PCR Inhibitor and ATP, Lot Traceable |
Storage Requirements | Ambient |
For Use With (Application) | PCR and Real Time PCR, viscous samples |
Sterilization Method | Electron Beam |
Features | Low Retention Technology, Ultra Clear Resin |
Compatibility | Universal |
Format | Hinged Rack |
Tip Style | Low Retention,Extended Length,Micro Point,Finely Tapered,Conical |
Tip Style | Low Retention,Extended Length,Micro Point,Finely Tapered,Conical |
Tip Style | Low Retention,Extended Length,Micro Point,Finely Tapered,Conical |
Tip Style | Low Retention,Extended Length,Micro Point,Finely Tapered,Conical |
Tip Style | Low Retention,Extended Length,Micro Point,Finely Tapered,Conical |
Packaging | Filtered Rack Sterile |
Quantity | 960 |
Product Type | Pipette Tip |
Color Code | Clear |
Disposable | Single-use |
For Use With (Equipment) | Universal Fit |
Graduated | Yes |
Product Line | SureOne |
Material | Polypropylene |
Shape | Conical |
Graduations | 10 μL, 50 μL, 100 μL, 200 μL |