Use promo code "24135" to get your promotional price.
Sartorius SafetySpace™ Extended Filter Tip
SafetySpace™ tips have extra space between the sample and the filter, which protects the sample and the pipette from cross-contamination.
152.99 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Use promo code "24135" to get your promotional price.
Additional Details:
Weight:
1.74000kg
Specification
Description | Free of DNase, Rnase, and endotoxins |
---|---|
Volume (Metric) | 0.5 to 200 μL |
Filtered | Filtered |
Sterility | Sterile |
Color | Colorless |
Autoclavable | Not Autoclavable |
Length (English) | 3.05 in. |
Length (Metric) | 77.5 mm |
Certifications/Compliance | Manufactured according to ISO9001, ISO14001, and ISO13485; ISO class 8 clean room condition; CE complaint |
For Use With (Application) | Precision Liquid Dispensing |
Sterilization Method | E-Beam Pre-Sterilized |
Format | Rack |
Tip Style | Extended Length |
Packaging | Single Tray |
Product Type | Tips |
Disposable | Single-use |
For Use With (Equipment) | Sartorius |
Graduated | No |
Product Line | SafetySpace |
Material | Polypropylene |