2.30 GBP valid until 2025-07-22
Use promo code "24098" to get your promotional price.
Use promo code "24098" to get your promotional price.
Certifications/Compliance | Certified to ANSI Z87.1-2015 and EN 166; Markings: ANSI - Z87+, CE - CE EN166 1B |
---|---|
Lens Coating | Anti-fog,Scratch-resistant |
Lens Coating | Anti-fog,Scratch-resistant |
Color | Clear |
Certifications/Compliance (Europe) | CE EN 166 ANSI Z87.1 |
Dimensions (L x W x H) | 17 x 9 x 8 cm |
Frame Color | Clear |
Headband Material | Elastic |
Size | One size |
Lens Material | Polycarbonate |
Frame Material | PVC |
UV Protection | Yes |