Use promo code "24075" to get your promotional price.
BRAND™ accu-jet™ S, with Universal Power Supply
The accu-jet™ S with universal AC adapter brings versatility and endurance to your team.
375.69 GBP valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Use promo code "24075" to get your promotional price.
Additional Details:
Weight:
0.20500kg
Specification
Battery Type | 2 NiMH/AAA |
---|---|
Autoclavable | Autoclavable |
Color | Amethyst |
Disposable | Reusable |
Grip Type | Soft Push Button |
Material | Polycarbonate, PBT |
Number of Channels | 1 |
Sterility | Non-sterile |
Type | Pipette Controller |
Volume (Metric) | 3214.80 mL |
Filter Type | 0.2 μm PTFE |
Includes | Pipette controller, nickel-metal hydride battery, power adapter, 2 x membrane filters, wall mount, operating manual |
Temperature (Metric) Operating | 10°C to 40°C |
Warranty | 12 Months |
Length (Metric) | 23.9 cm |
Width (Metric) | 23 cm |
Height (Metric) | 6.2 cm |
Weight (Metric) | 639 g |