Fisherbrand™ Traceable™ Refrigerator/Freezer Digital Thermometer
Reliably monitor and be alerted to any issues affecting the contents of your fridge or freezer without even opening the door using the Traceable™ Refrigerator/Freezer Digital Thermometer.
Additional Details:
Weight:
0.22680kg
Specification
| Certifications/Compliance | NIST |
|---|---|
| Depth (English) | 0.75 in. |
| Depth (Metric) | 19 mm |
| Diameter (English) Probe | 0.1875 in. |
| Diameter (Metric) Probe | 4.7625 mm |
| For Use With (Application) | Temperature Monitoring |
| Height (English) | 4.25 in. |
| Height (Metric) | 107.95 mm |
| Includes | Thermometer, bottle probe, hook and loop tape, AAA batteries, quick start guide, and Traceable™ certificate |
| Length (English) Cable | 120 in. |
| Length (English) Probe | 7/8 in. |
| Length (Metric) | 21 cm |
| Length (Metric) Cable | 304.8 cm |
| Length (Metric) Probe | 22.225 mm |
| Material | Plastic |
| Probe Type | Bottle |
| Resolution | 1° |
| Product Type | Digital thermometer |
| Description | Fridge/Freezer Digital Thermometer with Bottle Probe |
| Max. Temperature (English) | 158°F |
| Max. Temperature (Metric) | 70°C |
| Min. Temperature (English) | -58°F |
| Min. Temperature (Metric) | -50°C |
| Temperature Range (English) | -58° to +158°F |
| Temperature Range (Metric) | -50°C to +70°C |
| Calibrated | Yes |
| For Use With (Equipment) | Baths, Incubators, Refrigerators, or Freezers |
| Battery Type | AAA |
| Accuracy | ±1°C |
| Display | LCD |
| Width (Metric) | 11 cm |