Fisherbrand™ Safety Bottle Carrier
The Safety Bottle Carrier provides a safe and reliable way to transport a variety of bottled reagents in your lab.
Overview
73.79 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Additional Details:
Additional Details: Weight: 0.90000kg
Product Code. 17830782
Quantity Price
1 £ 92.5 / Each
Estimated Shipment
16-07-2025
Add to basket
  • Description and Specification
  • Accessories and Spare Parts
Description and Specification
Specification
Includes 1 Safety Bottle Carrier vessel and attachable handle
Product Type Bottle Holder
Description Safety Bottle Carrier, 4.5 L, Black
Array 1 x 1
Autoclavable Not Autoclavable
Color Black
Holds Bottles
Material Polypropylene
Positions 1
Shape Cylindrical
Shelves None
Stackable No
Sterility Non-sterile
For Use With (Application) Transportation of bottled regeants
For Use With (Equipment) 4.5 L bottles
Size 4.5 L
Temperature (Metric) Range -20 to 60°C
Length (Metric) 18.3 cm
Width (Metric) 18.3 cm
Height (Metric) 26.7 cm (40.6 cm Height, Handle Extended)
Diameter (Metric) Opening 18.3 cm
Diameter (Metric) Inner 18.3 cm
Weight (Metric) .25 kg
  • Robust Design
  • Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
  • Handle can be pressed down to either side for easier loading and unloading
  • Centering ring stabilizes bottle in the center of carrier for safer, steadier transport
  • Carriers come in four colors with designated labeling areas for easy identification
  • Made of chemical resistant thermoplastic elastomer - PP blend
Accessories and Spare Parts