Ecolab™ Klercide Sporicidal Low Residue Peroxide Blended with WFI Capped
Klercide Sporicidal Low Residue Peroxide is a very low residue blend of high grade 6% hydrogen peroxide and water for injection. The biocide blend is 0.2 μm filtered and sterile filled under Grade A laminar flow, then triple bagged in a Grade B cleanroom.
Overview
613.60 GBP valid until 2025-06-21
Use promo code "24097" to get your promotional price.
Additional Details:
Additional Details: CAS Number: 7722-84-1 Weight: 20.43000kg
Product Code. 17874554
Quantity Price
1 £ 1107.6 / 20 L
In stock, ready to ship: 23
Add to basket
  • Description and Specification
  • Alternative Products
  • Safety and Handling
  • Special Offers
Description and Specification
Specification
Product Type Cleanroom Biocide
Form Clear Liquid
Scent Unscented
Sterility Sterile
Appearance Clear
Composition 6% Hydrogen Peroxide and Deionized Water
Container Type Capped Bottle
Disposable Single-use
Filtered Filtered
Flammable No
Quantity 4 x 5 L
Temperature Range (English) 0°C to 25°C
  • BPR approved
  • Endotoxin level guaranteed to be less than 0.25 EU/mL
  • Broad spectrum activity that includes proven effects against bacterial spores
  • Can be used in rotation with any other Ecolab™ biocide or to pro-actively reduce bioburden levels including bacterial spores
  • Non-corrosive and triple bagged
  • 100% compatible with gassing isolators
  • Suitable for use on all types of hard surfaces in cleanrooms and controlled areas
  • Very low residue profile, reducing the need to perform rinsing steps after application
  • 5 L capped container for large surfaces
  • Certificates of Analysis and Sterility are available for every batch
  • Full supporting technical documentation is available
Alternative Products
Safety and Handling
[Klercide Sporicidal Low Residue Peroxide]
[Warning]
[{code=EYE DAM. 2, sequence=1, description=Serious eye damage/eye irritation Category 2}]
[{code=H319, sequence=1, description=Causes serious eye irritation.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Hydrogen peroxide}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]
Product Identifier
  • Klercide Sporicidal Low Residue Peroxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319- Causes serious eye irritation.
Precautionary Statement
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental Information
  • MIXTURE LIST- Contains : Hydrogen peroxide
Special Offers