Use promo code "25339" to get your promotional price.
Thermo Scientific™ SureSTART™ 9 mm Screw Caps, Level 2 High-throughput Applications
Use Level 2 Thermo Scientific™ SureSTART™ 9 mm Screw Caps with screw vials that have a 9 mm opening for your high-throughput applications, when robustness and reproducibility are key.
19.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Additional Details:
Weight:
0.09000kg
Specification
Color | Green |
---|---|
Material | Polypropylene |
Pre-slit | No |
Septum | Red Silicone/Ivory PTFE |
Hardness | 45 Shore |
Product Line | SureSTART Level 2 |
Quantity | 100 Pk. |
Thickness (Metric) | 1 mm |
Certifications/Compliance | Specification Certified |
Type | AVCS Screw Cap |
Closure Size | 9 mm |
For Use With (Equipment) | Screw Thread Vial |